Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02027239_s1
          See other MAP2K3OS GE Assays ›
          Gene Symbol:
          Map2k3os
          Gene Name:
          mitogen-activated protein kinase kinase 3, opposite strand
          Gene Aliases:
          F3-1, Gm16516, Gtlf3a
          Chromosome Location:
          Chr.11: 60920941 - 60931867 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02027239_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Map2k3os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027800.1 2 1183 157
          GenBank mRNA AK009371.1 - 2 1184 157
          AK010055.1 - 2 1184 157
          AK010107.1 - 2 1184 157
          BC031537.1 - 2 1172 157

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 BC031537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 AK010055.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 AK010107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 NR_027800.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 AK009371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 BC031537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 AK010055.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 AK010107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 NR_027800.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P153013 AK009371.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600946 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707620 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76610 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137263 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127978 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 BC031537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 AK010055.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 AK010107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 NR_027800.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 AK009371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 BC031537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 AK010055.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 AK010107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 NR_027800.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P153013 AK009371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 BC031537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 AK010055.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 AK010107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 NR_027800.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 AK009371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 BC031537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 AK010055.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 AK010107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 NR_027800.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P153013 AK009371.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10386635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386634 - -
          Affymetrix HT MG-430 PM Array Plate 1419203_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10386633 - -
          Mouse Genome 430 2.0 Array 1419203_at - -
          Mouse Genome 430 2.0 Array 1419203_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100026313.mm.1 NR_027800.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026313.mm.1 NR_027800.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026313.mm.1 TC1100002902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100026316.mm.1 NR_027800.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026316.mm.1 NR_027800.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026316.mm.1 TC1100002902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100026312.mm.1 NR_027800.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026312.mm.1 NR_027800.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026312.mm.1 TC1100002902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100026311.mm.1 NR_027800.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026311.mm.1 NR_027800.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100026311.mm.1 TC1100002902.mm.1 MTA TC
          Mouse Expression Set 430 1419203_at - -
          Mouse Expression Set 430 1419203_PM_at - -
          Murine Genome U74v2 Set 165922_PM_at - -
          Murine Genome U74v2 Set 165922_at - -
          Mouse Genome 430A 2.0 Array 1419203_PM_at - -
          Mouse Genome 430A 2.0 Array 1419203_at - -
          Affymetrix HT Mouse Genome 430A Array 1419203_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419203_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 924673 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline