Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02029600_s1
          See other OLFR722 GE Assays ›
          Gene Symbol:
          Olfr722
          Gene Name:
          olfactory receptor 722
          Gene Aliases:
          MOR241-3
          Chromosome Location:
          Chr.14: 49894258 - 49901941 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02029600_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr722
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146494.2 NP_666705.1 2 912 134
          GenBank mRNA BC051403.1 - 2 912 6392119 134

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr722

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963324 BC051403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963324 NM_146494.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923440 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102729 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109380 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963324 BC051403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963324 NM_146494.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P406931 BC051403.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P406931 NM_146494.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29595992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596012 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08108176 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109380 - -
          Whole Mouse Genome G4122A A_51_P406931 BC051403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406931 NM_146494.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135528 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109380 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11446184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11557849 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11446191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11446200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29595971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11533815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11160346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11446185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11313579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29595987 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10419428 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419427 - -
          Affymetrix Mouse Gene 1.1 ST Array 10419426 - -
          Affymetrix Mouse Clariom S Array PSR1400014818.mm.1 NM_146494.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014818.mm.1 NM_146494.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014818.mm.1 TC1400001969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400014819.mm.1 NM_146494.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014819.mm.1 NM_146494.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014819.mm.1 TC1400001969.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 507667 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1170138 - -
          Illumina mouse-6 v1.0 expression beadchip 1170138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170138 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2632903 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2632903 - -
          mouse-6 v1.1 (Illumina) scl45625.2.7_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45625.2.7_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2893483 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2632903 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2632903 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2893483 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0