Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02029641_u1
          See other DEFB11 GE Assays ›
          Gene Symbol:
          Defb11
          Gene Name:
          defensin beta 11
          Gene Aliases:
          BD-11, Defb8
          Chromosome Location:
          Chr.8: 21905374 - 21906432 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02029641_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139221.2 NP_631967.1 1 91 154
          GenBank mRNA AJ437646.1 - 1 70 154
          BC099849.1 - 1 59 40038952 154
          CJ044726.1 - 1 92 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001856.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defb11

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          defense response to bacterium
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317254 NM_139221.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317254 AJ437646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317254 BC099849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317254 CJ044726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317254 NM_139221.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317254 AJ437646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317254 BC099849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317254 CJ044726.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107663 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317254 NM_139221.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317254 AJ437646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317254 BC099849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317254 CJ044726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317254 NM_139221.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317254 AJ437646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317254 BC099849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317254 CJ044726.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25708224 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107663 - -
          Whole Mouse Genome G4122A A_51_P317254 NM_139221.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317254 AJ437646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317254 BC099849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317254 CJ044726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317254 NM_139221.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317254 AJ437646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317254 BC099849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317254 CJ044726.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134071 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107663 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07320321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25708153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07393410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06976107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06976105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25708190 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577445 - -
          Affymetrix HT MG-430 PM Array Plate 1452546_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10577444 - -
          Mouse Genome 430 2.0 Array 1452546_PM_x_at - -
          Mouse Genome 430 2.0 Array 1452546_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246081_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246081_at - -
          Affymetrix Mouse Clariom S Array PSR0800014629.mm.1 NM_139221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014629.mm.1 NM_139221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014629.mm.1 TC0800001856.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014627.mm.1 NM_139221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014627.mm.1 NM_139221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014627.mm.1 TC0800001856.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014628.mm.1 NM_139221.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014628.mm.1 NM_139221.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014628.mm.1 TC0800001856.mm.1 MTA TC
          Mouse Expression Set 430 1452546_x_at - -
          Mouse Expression Set 430 1452546_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1452546_x_at - -
          Mouse Genome 430A 2.0 Array 1452546_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1452546_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1452546_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 662659 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2370440 - -
          Illumina mouse-6 v1.0 expression beadchip 2370440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251369 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251369 - -
          mouse-6 v1.1 (Illumina) scl35030.3.1_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35030.3.1_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599861 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599861 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35030.3.1_41-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0