Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02342844_g1
          See other SPEER4F GE Assays ›
          Gene Symbol:
          Speer4f
          Gene Name:
          spermatogenesis associated glutamate (E)-rich protein 4f
          Gene Aliases:
          4922502J04Rik, SPEER-4F
          Chromosome Location:
          Chr.5: 17476098 - 17480936 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02342844_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Speer4f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027609.2 NP_081885.1 4 - 5 607 133
          GenBank mRNA AK015022.1 - 4 - 5 610 133
          BC116901.1 - 4 - 5 633 40126220 133
          BC116903.1 - 4 - 5 633 40126222 133

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Speer4f

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056403 AK015022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056403 BC116901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056403 BC116903.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056403 NM_027609.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056403 AK015022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056403 BC116901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056403 BC116903.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056403 NM_027609.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249073 AK015022.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249073 NM_027609.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23505392 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491478 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491516 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491578 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156567 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491636 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491614 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491588 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491562 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491553 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116897 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05491603 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01044921 - -
          Whole Mouse Genome G4122A A_51_P249073 AK015022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249073 NM_027609.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120299 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116897 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05491636 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156567 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05491608 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05491562 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01044921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23505932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23505963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04110800 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10519883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519881 - -
          Affymetrix HT MG-430 PM Array Plate 1420791_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10519879 - -
          Mouse Genome 430 2.0 Array 1420791_x_at - -
          Mouse Genome 430 2.0 Array 1420791_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70935_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70935_PM_at - -
          Mouse Expression Set 430 1420791_x_at - -
          Mouse Expression Set 430 1420791_PM_x_at - -
          Murine Genome U74v2 Set 170588_at - -
          Murine Genome U74v2 Set 170588_PM_at - -
          Murine Genome U74v2 Set 169952_PM_at - -
          Murine Genome U74v2 Set 169952_at - -
          Mouse Genome 430A 2.0 Array 1420791_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1420791_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420791_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420791_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 562313 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1500711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500711 - -
          mouse-6 v1.1 (Illumina) scl28086.4.1_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500711 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28086.4.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2842348 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2842348 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692573 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline