Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02343059_s1
          See other RNF113A1 GE Assays ›
          Gene Symbol:
          Rnf113a1
          Gene Name:
          ring finger protein 113A1
          Gene Aliases:
          2810428C21Rik, Rnf113a
          Chromosome Location:
          Chr.X: 37191343 - 37192467 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02343059_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf113a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153503.1 NP_705723.1 1 363 128
          GenBank mRNA AK013178.1 - 1 424 128
          AK146283.1 - 1 462 128
          BC024906.1 - 1 363 3495576 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf113a1

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32322125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32322124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32322126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32322123 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285487 NM_153503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285487 AK013178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285487 AK146283.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285487 BC024906.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285487 NM_153503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285487 AK013178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285487 AK146283.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P285487 BC024906.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55054 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120351 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285487 NM_153503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285487 AK013178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285487 AK146283.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285487 BC024906.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285487 NM_153503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285487 AK013178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285487 AK146283.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P285487 BC024906.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285487 NM_153503.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285487 AK013178.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285487 AK146283.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285487 BC024906.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285487 NM_153503.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285487 AK013178.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285487 AK146283.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285487 BC024906.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32322125 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120351 - -
          Whole Mouse Genome G4122A A_51_P285487 NM_153503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285487 AK013178.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285487 AK146283.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285487 BC024906.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285487 NM_153503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285487 AK013178.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285487 AK146283.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285487 BC024906.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127194 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120351 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14555205 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14555194 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14555208 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32322123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32322125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32322126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32322124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32322132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14918045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14644096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15199717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14858431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32322136 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10599245 - -
          Affymetrix HT MG-430 PM Array Plate 1424837_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10599243 - -
          Mouse Genome 430 2.0 Array 1424837_at - -
          Mouse Genome 430 2.0 Array 1424837_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69942_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69942_PM_at - -
          Mouse Expression Set 430 1424837_PM_at - -
          Mouse Expression Set 430 1424837_at - -
          Murine Genome U74v2 Set 170330_at - -
          Murine Genome U74v2 Set 170330_PM_at - -
          Murine Genome U74v2 Set 167363_f_at - -
          Murine Genome U74v2 Set 167363_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1424837_PM_at - -
          Mouse Genome 430A 2.0 Array 1424837_at - -
          Affymetrix HT Mouse Genome 430A Array 1424837_at - -
          Affymetrix HT Mouse Genome 430A Array 1424837_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 643952 - -
          ABI Mouse Genome Survey Microarray 841683 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2650520 - -
          Illumina mouse-6 v1.0 expression beadchip 2650520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002650520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237083 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237083 - -
          mouse-6 v1.1 (Illumina) scl54988.1.64_253-S - -
          Illumina Sentrix mouse-6.v1 bead array 2650520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54988.1.64_253-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2650520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2588728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2588728 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54988.1.64_253-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline