Search
Search
| Gene Symbol: Xkr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011873.2 | NP_001011873.1 | 3 - 4 | 953 | 176 | |
| GenBank mRNA | AY534255.1 | - | 3 - 4 | 953 | 176 | |
| BC147699.1 | - | 2 - 3 | 559 | 9056672 | 176 | |
| BC158121.1 | - | 2 - 3 | 814 | 9088194 | 176 | |
Set Membership: |
|