Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02343755_g1
          See other KLK1B22 GE Assays ›
          Gene Symbol:
          Klk1b22
          Gene Name:
          kallikrein 1-related peptidase b22
          Gene Aliases:
          Egfbp-1, Egfbp1, Klk22, mGk-22
          Chromosome Location:
          Chr.7: 44112683 - 44116876 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02343755_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klk1b22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010114.1 NP_034244.1 3 - 4 509 162
          GenBank mRNA BC106155.1 - 3 - 4 513 4189167 162

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000790.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klk1b22

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          zymogen activation
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112986 NM_010114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112986 BC106155.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153745 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112986 NM_010114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112986 BC106155.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25085740 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01505591 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05887337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25085767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06400128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06572842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06117664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06565768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05887330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25085733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552596 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552594 - -
          Mouse Genome 430 2.0 Array 1425182_PM_x_at - -
          Mouse Genome 430 2.0 Array 1425182_x_at - -
          Affymetrix Mouse Clariom S Array PSR0700006415.mm.1 TC0700000790.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006415.mm.1 NM_010114.2 Refseq
          Mouse Expression Set 430 1425182_PM_x_at - -
          Mouse Expression Set 430 1425182_x_at - -
          Murine Genome U74v2 Set 101289_PM_f_at - -
          Murine Genome U74v2 Set 101289_f_at - -
          Mouse Genome 430A 2.0 Array 1425182_x_at - -
          Mouse Genome 430A 2.0 Array 1425182_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2946653 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2946653 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline