Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02344116_g1
          See other GM5444 GE Assays ›
          Gene Symbol:
          Gm5444
          Gene Name:
          predicted gene 5444
          Gene Aliases:
          EG432723
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02344116_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5444
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK030184.1 - 2 - 3 174 104

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998335 AK030184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123756 AK030184.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10793234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10719504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11046884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10871516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10755647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10646713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10956344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10755650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10979424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10755638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11093589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10662802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10625900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28793927 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 467630 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106860164 - -
          Illumina mouse-6 v1.0 expression beadchip 106860164 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223676 - -
          Illumina Sentrix mouse-6.v1 bead array 106860164 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106860164 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2946134 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline