Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02344321_s1
          See other OLFR1461 GE Assays ›
          Gene Symbol:
          Olfr1461
          Gene Name:
          olfactory receptor 1461
          Gene Aliases:
          MOR202-35
          Chromosome Location:
          Chr.19: 13163098 - 13165954 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02344321_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1461
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146302.1 NP_666414.1 1 68 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1461

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065804 NM_146302.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179975 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167395 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125378 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065804 NM_146302.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963780 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167395 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125378 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179975 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126645 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125378 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179975 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167395 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14360074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14389719 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14389723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14332274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14441259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14441257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14379761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14360075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14441263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963760 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461795 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461794 - -
          Affymetrix Mouse Clariom S Array PSR1900003030.mm.1 NM_146302.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003030.mm.1 NM_146302.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003030.mm.1 TC1900000266.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2964937 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2964937 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline