| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100005933.mm.1 |
NM_172849.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005933.mm.1 |
NM_172849.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005933.mm.1 |
TC0100000697.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005929.mm.1 |
NM_172849.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005929.mm.1 |
NM_172849.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005929.mm.1 |
TC0100000697.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005930.mm.1 |
NM_172849.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005930.mm.1 |
NM_172849.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005930.mm.1 |
TC0100000697.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005931.mm.1 |
NM_172849.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005931.mm.1 |
NM_172849.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005931.mm.1 |
TC0100000697.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005932.mm.1 |
NM_172849.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005932.mm.1 |
NM_172849.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005932.mm.1 |
TC0100000697.mm.1 |
MTA TC |