| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10413218 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10413217 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10413219 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1454280_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1432572_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10413216 |
- |
- |
| Mouse Genome 430 2.0 Array |
1432572_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1454280_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1432572_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1454280_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
77592_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
77592_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1400001807.mm.1 |
NR_033492.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001807.mm.1 |
NR_033492.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001807.mm.1 |
TC1400000244.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400001806.mm.1 |
NR_033492.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001806.mm.1 |
NR_033492.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001806.mm.1 |
TC1400000244.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400001808.mm.1 |
NR_033492.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001808.mm.1 |
NR_033492.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001808.mm.1 |
TC1400000244.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1454280_at |
- |
- |
| Mouse Expression Set 430 |
1432572_PM_at |
- |
- |
| Mouse Expression Set 430 |
1432572_at |
- |
- |
| Mouse Expression Set 430 |
1454280_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1454280_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1432572_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1454280_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1432572_PM_at |
- |
- |