Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02344527_s1
          See other GPR150 GE Assays ›
          Gene Symbol:
          Gpr150
          Gene Name:
          G protein-coupled receptor 150
          Gene Aliases:
          C030001A19Rik, PGR11
          Chromosome Location:
          Chr.13: 76054851 - 76056996 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02344527_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr150
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175495.2 NP_780704.1 1 2090 148
          GenBank mRNA AK047609.1 - 1 2090 148

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr150

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          biological_process
          molecular_function
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152745 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152749 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152746 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152747 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152752 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152748 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479548 NM_175495.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479548 AK047609.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479548 NM_175495.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479548 AK047609.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89772 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104749 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479548 NM_175495.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479548 AK047609.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479548 NM_175495.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479548 AK047609.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479548 NM_175495.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479548 AK047609.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479548 NM_175495.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479548 AK047609.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29152741 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104749 - -
          Whole Mouse Genome G4122A A_52_P624685 BC118612.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624685 BC118614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624685 NM_175495.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624685 AK047609.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624685 AY255541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479548 NM_175495.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479548 AK047609.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479548 NM_175495.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479548 AK047609.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136060 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104749 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10943156 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10943159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10943166 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10943167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10943138 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152746 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152748 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152745 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152747 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152752 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29152749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29152742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10747761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10472544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10589630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10943151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10643891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10889882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10943141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10472542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10943157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29152768 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10410719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410720 - -
          Affymetrix HT MG-430 PM Array Plate 1447485_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442261_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10410716 - -
          Mouse Genome 430 2.0 Array 1447485_at - -
          Mouse Genome 430 2.0 Array 1442261_at - -
          Mouse Genome 430 2.0 Array 1442261_PM_at - -
          Mouse Genome 430 2.0 Array 1447485_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 238725_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 238725_at - -
          Mouse Expression Set 430 1442261_at - -
          Mouse Expression Set 430 1442261_PM_at - -
          Mouse Expression Set 430 1447485_at - -
          Mouse Expression Set 430 1447485_PM_at - -
          Murine Genome U74v2 Set 136115_at - -
          Murine Genome U74v2 Set 136115_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447485_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442261_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447485_at - -
          Affymetrix HT Mouse Genome 430B Array 1442261_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 552433 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2570440 - -
          Illumina mouse-6 v1.0 expression beadchip 2570440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692211 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692211 - -
          mouse-6 v1.1 (Illumina) scl43744.2.395_111-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43744.2.395_111-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2985788 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2985788 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline