Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02344630_s1
          See other OLFR1344 GE Assays ›
          Gene Symbol:
          Olfr1344
          Gene Name:
          olfactory receptor 1344
          Gene Aliases:
          4930415J05Rik, MOR124-1
          Chromosome Location:
          Chr.7: 6439902 - 6440867 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02344630_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1344
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177061.3 NP_796035.2 1 838 89
          GenBank mRNA BC147050.1 - 1 904 9056072 89
          BC147051.1 - 1 904 9056049 89

          Back To Top

          More Information


          Additional Information:

          This assay targets the same location as MTA transcript cluster(s) TC0700000079.mm
          This assay targets the same location as RTA transcript cluster(s) TC0100004751.rn

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1344

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012141 BC147050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012141 BC147051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012141 NM_177061.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012141 BC147050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012141 BC147051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012141 NM_177061.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P570487 BC147050.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P570487 BC147051.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P570487 NM_177061.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24955474 - -
          Whole Mouse Genome G4122A A_52_P570487 BC147050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P570487 BC147051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P570487 NM_177061.3 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24955473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06569721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06337327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06337324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06505321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06569718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05892924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24955476 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549867 - -
          Affymetrix HT MG-430 PM Array Plate 1441735_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10549862 - -
          Mouse Genome 430 2.0 Array 1441735_at - -
          Mouse Genome 430 2.0 Array 1441735_PM_at - -
          Mouse Genome 430 2.0 Array 1459717_at - -
          Mouse Genome 430 2.0 Array 1459717_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700000893.mm.1 TC0700000079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000892.mm.1 TC0700000079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000892.mm.1 NM_177061.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000892.mm.1 NM_177061.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000890.mm.1 TC0700000079.mm.1 MTA TC
          Mouse Expression Set 430 1459717_PM_at - -
          Mouse Expression Set 430 1441735_PM_at - -
          Mouse Expression Set 430 1459717_at - -
          Mouse Expression Set 430 1441735_at - -
          Murine Genome U74v2 Set 138187_at - -
          Murine Genome U74v2 Set 138187_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441735_at - -
          Affymetrix HT Mouse Genome 430B Array 1441735_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459717_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459717_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3780739 - -
          Illumina mouse-6 v1.0 expression beadchip 3780739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2748047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160374 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160374 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2748047 - -
          mouse-6 v1.1 (Illumina) scl33102.9.1_233-S - -
          mouse-6 v1.1 (Illumina) GI_59676556-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33102.9.1_233-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160374 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2748047 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160374 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline