Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02345012_s1
          See other OLFR1173 GE Assays ›
          Gene Symbol:
          Olfr1173
          Gene Name:
          olfactory receptor 1173
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 88274109 - 88275047 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02345012_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1173
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207566.1 NP_997449.1 1 461 126
          GenBank mRNA BC104085.2 - 1 461 40042097 126
          BC104278.1 - 1 461 40041579 126
          BC104279.1 - 1 461 40041581 126
          BC127020.1 - 1 461 40042733 126
          BC134355.1 - 1 461 40041582 126

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1173

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048009 BC134355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048009 NM_207566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048009 BC104085.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048009 BC104278.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048009 BC104279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048009 BC134355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048009 NM_207566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048009 BC104085.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048009 BC104278.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048009 BC104279.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01357227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01559273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02037584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01177644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01559280 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484732 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484731 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3610546 - -
          Illumina mouse-6 v1.0 expression beadchip 3610546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252315 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252315 - -
          mouse-6 v1.1 (Illumina) scl0404329.1_55-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610546 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0404329.1_55-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252315 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline