Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02345037_g1
          See other OLFR1321 GE Assays ›
          Gene Symbol:
          Olfr1321
          Gene Name:
          olfactory receptor 1321
          Gene Aliases:
          MOR264-26
          Chromosome Location:
          Chr.X: 49726973 - 49727932 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02345037_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1321
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207631.1 NP_997514.1 1 913 134
          GenBank mRNA BC119046.1 - 1 980 8733795 134
          BC119381.1 - 1 980 8734130 134

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1321

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123271 NM_207631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123271 BC119046.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123271 BC119381.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511212 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123271 NM_207631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123271 BC119046.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123271 BC119381.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P421177 NM_207631.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P421177 BC119046.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P421177 BC119381.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32369862 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03670676 - -
          Whole Mouse Genome G4122A A_51_P421177 NM_207631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P421177 BC119046.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P421177 BC119381.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03670676 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14761718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14524635 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14524633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14761712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32369884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14607973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14761723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14771978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15049069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14604592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14923494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15049080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14771980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14761717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14524634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15185270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14887875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32369911 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10604471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604472 - -
          Affymetrix Mouse Gene 1.1 ST Array 10604470 - -
          Affymetrix Mouse Clariom S Array PSR0X00013821.mm.1 TC0X00002176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00013822.mm.1 NM_207631.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00013822.mm.1 NM_207631.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00013822.mm.1 TC0X00002176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00013823.mm.1 TC0X00002176.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247217 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2877571 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2877571 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline