Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm02345133_m1
See other GSDMA3 GE Assays ›
Gene Symbol:
Gsdma3
Gene Name:
gasdermin A3
Gene Aliases:
Bsk, Dfl, Fgn, Gsdm1l, Gsdm3, Rco2, Rim3
Chromosome Location:
Chr.11: 98626360 - 98638308 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm02345133_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Gsdma3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001007461.1
NP_001007462.1
7 - 8
859
104
GenBank mRNA
AY679090.1
-
7 - 8
859
104
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC1100001575.mm.1
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Recommended Assay
Mus musculus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Gsdma3
Gene Ontology Categories:
Function(s)
Process(es)
hair follicle development
sebaceous gland cell differentiation
multicellular organism development
hair follicle morphogenesis
somatic stem cell population maintenance
avascular cornea development in camera-type eye
hair cycle
catagen
anagen
positive regulation of apoptotic process
skin development
positive regulation of extrinsic apoptotic signaling pathway
molecular_function
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR1100014594.mm.1
NM_001007461.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1100014594.mm.1
NM_001007461.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1100014594.mm.1
TC1100001575.mm.1
MTA TC
Affymetrix Mouse Clariom S Array
PSR1100014595.mm.1
NM_001007461.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1100014595.mm.1
NM_001007461.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1100014595.mm.1
TC1100001575.mm.1
MTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit