Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02345245_s1
          See other TAAR9 GE Assays ›
          Gene Symbol:
          Taar9
          Gene Name:
          trace amine-associated receptor 9
          Gene Aliases:
          Gm231, taR-9
          Chromosome Location:
          Chr.10: 24108488 - 24109534 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02345245_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taar9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010831.1 NP_001010831.1 1 840 93
          GenBank mRNA BC139084.1 - 1 889 8862106 93
          BC139085.1 - 1 888 8862107 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Taar9

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of chemical stimulus
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          trace-amine receptor activity
          signal transducer activity
          G-protein coupled receptor activity
          G-protein coupled amine receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978977 BC139085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978977 NM_001010831.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978977 BC139084.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149948 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978977 BC139085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978977 NM_001010831.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978977 BC139084.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P353631 BC139085.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P353631 NM_001010831.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P353631 AY255559.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P353631 BC139084.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006829 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149948 - -
          Whole Mouse Genome G4122A A_52_P353631 BC139085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353631 NM_001010831.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353631 AY255559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353631 BC139084.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08421917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08386731 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08386751 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08421921 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08421916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08407499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08528122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08386736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08386729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08948402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08386731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08528123 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10368288 - -
          Affymetrix Mouse Gene 1.1 ST Array 10368287 - -
          Affymetrix Mouse Clariom S Array PSR1000013560.mm.1 TC1000001886.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224966 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2884207 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2884207 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline