Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02345612_s1
          See other OLFR1181 GE Assays ›
          Gene Symbol:
          Olfr1181
          Gene Name:
          olfactory receptor 1181
          Gene Aliases:
          MOR225-9P
          Chromosome Location:
          Chr.2: 88423088 - 88426023 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02345612_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1181
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011816.1 NP_001011816.1 1 752 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1181

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140338 NM_001011816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140338 NM_001011816.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115376 NM_001011816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115376 NM_001011816.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01290136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01271887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484746 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484745 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 742685 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160858 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160858 - -
          mouse-6 v1.1 (Illumina) GI_58801397-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160858 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160858 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline