Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02374555_sH
          See other I830012O16RIK GE Assays ›
          Gene Symbol:
          I830012O16Rik
          Gene Name:
          RIKEN cDNA I830012O16 gene
          Gene Aliases:
          Ifit3b
          Chromosome Location:
          Chr.19: 34607539 - 34613409 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02374555_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: I830012O16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005858.3 NP_001005858.2 1656 135
          GenBank mRNA AK077243.1 - 1645 135

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene I830012O16Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972872 AK151442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972872 AK152620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972872 NM_001005858.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972872 AK077243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972872 AK150632.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P117933 NM_001005858.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P117933 AK077243.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901746 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972872 AK151442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972872 AK152620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972872 NM_001005858.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972872 AK077243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972872 AK150632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P117933 NM_001005858.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P117933 AK077243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663686 AK077243.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117933 NM_001005858.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117933 AK077243.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14274775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14155897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081228 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10462622 - -
          Affymetrix Mouse Gene 1.1 ST Array 10462621 - -
          Affymetrix Mouse Clariom S Array PSR1900004474.mm.1 NM_001005858.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004474.mm.1 NM_001005858.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004474.mm.1 TC1900000502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004472.mm.1 NM_001005858.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004472.mm.1 NM_001005858.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004472.mm.1 TC1900000502.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 888038 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246194 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246194 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline