Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02376544_g1
          See other TRIM30B GE Assays ›
          Gene Symbol:
          Trim30b
          Gene Name:
          tripartite motif-containing 30B
          Gene Aliases:
          A530023O14Rik, Trim30-1
          Chromosome Location:
          Chr.7: 104355398 - 104358646 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02376544_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trim30b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175648.2 NP_783579.1 2 650 107
          GenBank mRNA AK041488.1 - 2 650 107
          AK162946.1 - 3084 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003941.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trim30b

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2192961 AK162946.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2192961 AK041488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2192961 NM_175648.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151247 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177092 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143778 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2192961 AK162946.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2192961 AK041488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2192961 NM_175648.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387885 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143778 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177092 - -
          Whole Mouse Genome G4122A A_51_P274803 AK080092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P274803 AK040770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P274803 AK041488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P274803 NM_175648.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129331 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143778 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177092 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06275837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06048605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06074578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06275837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06558104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06494797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05993773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06074569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06048603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06507595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566353 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566352 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566351 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566354 - -
          Affymetrix HT MG-430 PM Array Plate 1444179_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10566350 - -
          Mouse Genome 430 2.0 Array 1444179_at - -
          Mouse Genome 430 2.0 Array 1444179_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 244183_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 244183_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700032939.mm.1 NM_175648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032939.mm.1 NM_175648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032939.mm.1 TC0700003941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032940.mm.1 NM_175648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032940.mm.1 NM_175648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032940.mm.1 TC0700003941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032943.mm.1 NM_175648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032943.mm.1 NM_175648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032943.mm.1 TC0700003941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032944.mm.1 NM_175648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032944.mm.1 NM_175648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032944.mm.1 TC0700003941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032942.mm.1 NM_175648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032942.mm.1 NM_175648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032942.mm.1 TC0700003941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032946.mm.1 NM_175648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032946.mm.1 NM_175648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032946.mm.1 TC0700003941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032947.mm.1 NM_175648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032947.mm.1 NM_175648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032947.mm.1 TC0700003941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032941.mm.1 NM_175648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032941.mm.1 NM_175648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032941.mm.1 TC0700003941.mm.1 MTA TC
          Mouse Expression Set 430 1444179_at - -
          Mouse Expression Set 430 1444179_PM_at - -
          Murine Genome U74v2 Set 134011_at - -
          Murine Genome U74v2 Set 134011_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444179_at - -
          Affymetrix HT Mouse Genome 430B Array 1444179_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 824819 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1230711 - -
          MouseWG-6 v2 Expression BeadChip 940139 - -
          Illumina mouse-6 v1.0 expression beadchip 50300 - -
          Illumina mouse-6 v1.0 expression beadchip 940139 - -
          Illumina mouse-6 v1.0 expression beadchip 1230711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2761994 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689647 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234137 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2761994 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689647 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234137 - -
          mouse-6 v1.1 (Illumina) scl0244183.3_17-S - -
          mouse-6 v1.1 (Illumina) scl0244183.1_172-S - -
          mouse-6 v1.1 (Illumina) scl00244183.1_117-S - -
          Illumina Sentrix mouse-6.v1 bead array 1230711 - -
          Illumina Sentrix mouse-6.v1 bead array 50300 - -
          Illumina Sentrix mouse-6.v1 bead array 940139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0244183.3_17-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0244183.1_172-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00244183.1_117-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940139 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50300 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2689651 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234137 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2761994 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234137 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2689651 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline