Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02376666_s1
          See other PRR18 GE Assays ›
          Gene Symbol:
          Prr18
          Gene Name:
          proline rich 18
          Gene Aliases:
          9630019K15Rik
          Chromosome Location:
          Chr.17: 8340406 - 8344113 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02376666_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prr18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178774.4 NP_848889.3 2 2862 94
          NR_028280.1 2 3018 94
          GenBank mRNA AK042857.1 - 1 632 94
          BC060190.1 - 2 3018 6833275 94

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prr18

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957128 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957133 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957132 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957130 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069251 BC060190.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069251 NM_178774.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069251 NR_028280.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511564 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123933 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069251 BC060190.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069251 NM_178774.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069251 NR_028280.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123933 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09553362 - -
          Whole Mouse Genome G4122A A_52_P494168 AK045379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494168 BC060190.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494168 AK035939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494168 AK042857.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494168 NM_178774.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106876 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123933 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957133 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957132 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957128 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957130 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957133 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10441658 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441659 - -
          Affymetrix HT MG-430 PM Array Plate 1435642_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455392_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10441657 - -
          Mouse Genome 430 2.0 Array 1455392_at - -
          Mouse Genome 430 2.0 Array 1455392_PM_at - -
          Mouse Genome 430 2.0 Array 1435642_at - -
          Mouse Genome 430 2.0 Array 1435642_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320111_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320111_at - -
          Affymetrix Mouse Clariom S Array PSR1700000749.mm.1 NM_178774.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000749.mm.1 NM_178774.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000749.mm.1 TC1700000090.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000748.mm.1 NR_028280.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000748.mm.1 NR_028280.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000748.mm.1 TC1700000090.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000751.mm.1 NM_178774.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000751.mm.1 NM_178774.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000751.mm.1 NR_028280.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000751.mm.1 NR_028280.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000751.mm.1 TC1700000090.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000752.mm.1 NM_178774.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000752.mm.1 NM_178774.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000752.mm.1 NR_028280.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000752.mm.1 NR_028280.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000752.mm.1 TC1700000090.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000750.mm.1 NM_178774.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000750.mm.1 NM_178774.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000750.mm.1 NR_028280.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000750.mm.1 NR_028280.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000750.mm.1 TC1700000090.mm.1 MTA TC
          Mouse Expression Set 430 1455392_PM_at - -
          Mouse Expression Set 430 1435642_at - -
          Mouse Expression Set 430 1455392_at - -
          Mouse Expression Set 430 1435642_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435642_at - -
          Affymetrix HT Mouse Genome 430B Array 1435642_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455392_at - -
          Affymetrix HT Mouse Genome 430B Array 1455392_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 305901 - -
          ABI Mouse Genome Survey Microarray 515398 - -
          ABI Mouse Genome Survey Microarray 632639 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2030064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030064 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2728682 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2728682 - -
          mouse-6 v1.1 (Illumina) scl51149.4_213-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51149.4_213-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2728682 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2728682 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51149.4_213-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline