Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02391416_s1
          See other OLFR222 GE Assays ›
          Gene Symbol:
          Olfr222
          Gene Name:
          olfactory receptor 222
          Gene Aliases:
          MOR256-45P
          Chromosome Location:
          Chr.11: 59570782 - 59571738 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02391416_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr222
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011789.1 NP_001011789.1 1 60 81
          GenBank mRNA BC147220.1 - 1 135 9056253 81
          BC147221.1 - 1 135 9056247 81

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr222

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061919 NM_001011789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061919 BC147220.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061919 BC147221.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061919 NM_001011789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061919 BC147220.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061919 BC147221.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09464002 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09463999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09506650 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09464000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09506649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08987375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09010739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09396492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09506652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09506650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09506647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09464001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09506649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837666 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10386417 - -
          Affymetrix Mouse Gene 1.1 ST Array 10386416 - -
          Affymetrix Mouse Clariom S Array PSR1100025909.mm.1 TC1100002855.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025911.mm.1 TC1100002855.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025910.mm.1 NM_001011789.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025910.mm.1 NM_001011789.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025910.mm.1 TC1100002855.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3005977 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3005977 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline