Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02391627_s1
          See other OLFR965 GE Assays ›
          Gene Symbol:
          Olfr965
          Gene Name:
          olfactory receptor 965
          Gene Aliases:
          MOR171-28
          Chromosome Location:
          Chr.9: 39719229 - 39720167 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02391627_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr965
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011859.1 NP_001011859.1 1 561 114

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr965

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2164764 NM_001011859.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958758 NM_001011859.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2164764 NM_001011859.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958758 NM_001011859.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439902 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07674042 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07742996 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07674053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08100408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07589954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07712468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08000644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08000658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08000645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08169813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07629139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584503 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584502 - -
          Affymetrix Mouse Clariom S Array PSR0900002907.mm.1 NM_001011859.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002907.mm.1 NM_001011859.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002907.mm.1 TC0900000434.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161298 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161298 - -
          mouse-6 v1.1 (Illumina) GI_58801475-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161298 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161298 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0