Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02391976_m1
          See other MPTX1 GE Assays ›
          Gene Symbol:
          Mptx1
          Gene Name:
          mucosal pentraxin 1
          Gene Aliases:
          1810030J14Rik, AV054416, Mptx
          Chromosome Location:
          Chr.1: 174330518 - 174332877 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02391976_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mptx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025470.3 NP_079746.1 1 - 2 119 91
          GenBank mRNA AK007658.1 - 1 - 2 120 91
          AK007963.1 - 1 - 2 119 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001646.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mptx1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057189 BC024348.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092633 AK007658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092633 AK007963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092633 NM_025470.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113002 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092633 AK007658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092633 AK007963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092633 NM_025470.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057189 BC024348.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20841908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20841875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20841907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20841897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20841878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20841880 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113002 - -
          Whole Mouse Genome G4122A A_51_P334930 AK007658.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334930 AK007963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334930 NM_025470.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100613 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109060 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113002 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00120051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20841926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00553240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00491595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00875691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00120051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20841874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00606640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00425203 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351960 - -
          Affymetrix HT MG-430 PM Array Plate 1417735_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351959 - -
          Mouse Genome 430 2.0 Array 1417735_PM_at - -
          Mouse Genome 430 2.0 Array 1417735_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66289_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66289_at - -
          Affymetrix Mouse Clariom S Array PSR0100013683.mm.1 NM_025470.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013683.mm.1 NM_025470.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013683.mm.1 TC0100001646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013684.mm.1 NM_025470.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013684.mm.1 NM_025470.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013684.mm.1 TC0100001646.mm.1 MTA TC
          Mouse Expression Set 430 1417735_PM_at - -
          Mouse Expression Set 430 1417735_at - -
          Murine Genome U74v2 Set 101386_PM_at - -
          Murine Genome U74v2 Set 101386_at - -
          Mouse Genome 430A 2.0 Array 1417735_at - -
          Mouse Genome 430A 2.0 Array 1417735_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417735_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417735_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 332848 - -
          ABI Mouse Genome Survey Microarray 587330 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4050619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050619 - -
          mouse-6 v1.1 (Illumina) scl066289.2_187-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066289.2_187-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2980898 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225061 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2980898 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline