Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02392026_g1
          See other ATP5H GE Assays ›
          Gene Symbol:
          Atp5h
          Gene Name:
          ATP synthase, H+ transporting, mitochondrial F0 complex, subunit D
          Gene Aliases:
          0610009D10Rik
          Chromosome Location:
          Chr.11: 115415691 - 115419919 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02392026_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp5h
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027862.1 NP_082138.1 5 - 6 389 76
          GenBank mRNA AF354051.1 - 4 - 5 382 76
          AK002376.1 - 5 - 6 389 76
          AK088617.1 - 4 - 5 384 76
          BC016547.1 - 5 - 6 387 2631309 76
          BC081431.1 - 5 - 6 388 6408035 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp5h

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          ATP synthesis coupled proton transport
          proton transport
          ATP metabolic process
          hydrogen ion transmembrane transporter activity
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153352 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153394 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153392 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153393 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152304 NM_027862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152304 AK002376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152304 BC016547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152304 BC081431.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152305 NM_027862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152305 AF354051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152305 AK002376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152305 AK088617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152305 BC016547.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920874 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104582 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139160 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161700 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152304 NM_027862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152304 AK002376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152304 BC016547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152304 BC081431.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152305 NM_027862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152305 AF354051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152305 AK002376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152305 AK088617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152305 BC016547.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P640355 NM_027862.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P640355 AF354051.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P640355 AK002376.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P640355 AK088617.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P640355 BC016547.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264186 NM_027862.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264186 AF354051.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264186 AK002376.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264186 AK088617.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264186 BC016547.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153374 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104582 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139160 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161700 - -
          Whole Mouse Genome G4122A A_52_P640355 NM_027862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P640355 AF354051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P640355 AK002376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P640355 AK088617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P640355 BC016547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264186 NM_027862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264186 AF354051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264186 AK002376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264186 AK088617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264186 BC016547.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117414 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161700 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139160 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09198633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09198628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09198632 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153352 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153393 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153392 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09361688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09082572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09361685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09152514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09176192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09634409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09361693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09198632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09082578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153355 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10427460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359298 - -
          Affymetrix HT MG-430 PM Array Plate 1435112_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1423676_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10359297 - -
          Affymetrix Mouse Gene 1.1 ST Array 10427459 - -
          Affymetrix Mouse Gene 1.1 ST Array 10542520 - -
          Affymetrix Mouse Gene 1.1 ST Array 10392930 - -
          Mouse Genome 430 2.0 Array 1423676_PM_at - -
          Mouse Genome 430 2.0 Array 1423676_at - -
          Mouse Genome 430 2.0 Array 1435112_a_at - -
          Mouse Genome 430 2.0 Array 1435112_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR1100038238.mm.1 NM_027862.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038238.mm.1 NM_027862.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038238.mm.1 TC1100004071.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038236.mm.1 NM_027862.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038236.mm.1 NM_027862.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038236.mm.1 TC1100004071.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100038237.mm.1 NM_027862.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038237.mm.1 NM_027862.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038237.mm.1 TC1100004071.mm.1 MTA TC
          Mouse Expression Set 430 1435112_a_at - -
          Mouse Expression Set 430 1435112_PM_a_at - -
          Mouse Expression Set 430 1423676_PM_at - -
          Mouse Expression Set 430 1423676_at - -
          Mouse Genome 430A 2.0 Array 1435112_a_at - -
          Mouse Genome 430A 2.0 Array 1423676_PM_at - -
          Mouse Genome 430A 2.0 Array 1423676_at - -
          Mouse Genome 430A 2.0 Array 1435112_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423676_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435112_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1435112_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423676_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 898495 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670438 - -
          Illumina mouse-6 v1.0 expression beadchip 670438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670438 - -
          mouse-6 v1.1 (Illumina) scl071679.3_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 670438 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl071679.3_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3004441 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2602633 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3004441 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl071679.3_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0