Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02392101_m1
          See other GM15514 GE Assays ›
          Gene Symbol:
          Gm15514
          Gene Name:
          predicted gene 15514
          Gene Aliases:
          EG434225, OTTMUSG00000023094
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02392101_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm15514
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY512917.1 - 382 115

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118839 AY512917.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99070 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11000 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59511 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178916 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176558 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118839 AY512917.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06313752 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178916 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06313796 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01597770 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06313800 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06313779 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06313753 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176558 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178916 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176558 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06313779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05947259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06346694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06171880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06182023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06030019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06182028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06434669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06346695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06114004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05887636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05957521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05902379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06187716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06114012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06410599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06483759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398705 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10556066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10556064 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161699 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161699 - -
          mouse-6 v1.1 (Illumina) GI_65301152-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline