Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02392382_s1
          See other RBMXL2 GE Assays ›
          Gene Symbol:
          Rbmxl2
          Gene Name:
          RNA binding motif protein, X-linked-like 2
          Gene Aliases:
          1700012H05Rik, Hnrnpg-t
          Chromosome Location:
          Chr.7: 107209445 - 107210916 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02392382_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbmxl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029660.2 NP_083936.1 1 752 84
          GenBank mRNA AK005907.1 - 1 753 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbmxl2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398385 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398383 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398388 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398380 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398384 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027747 AK005907.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027747 NM_029660.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794910 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027747 AK005907.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027747 NM_029660.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P397537 AK005907.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P397537 NM_029660.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397537 AK005907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397537 NM_029660.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129757 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398388 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398384 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398380 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398385 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25398383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06458274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06408222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05886605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06530007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06571807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06387522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05979536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06387528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06355338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06387524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06239959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06539990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398377 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556060 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556059 - -
          Mouse Genome 430 2.0 Array 1429343_PM_at - -
          Mouse Genome 430 2.0 Array 1429343_at - -
          Affymetrix Mouse Clariom S Array PSR0700013183.mm.1 TC0700001574.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013183.mm.1 NM_029660.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013183.mm.1 NM_029660.2 Refseq
          Mouse Expression Set 430 1429343_PM_at - -
          Mouse Expression Set 430 1429343_at - -
          Murine Genome U74v2 Set 165500_PM_at - -
          Murine Genome U74v2 Set 165500_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 347990 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249885 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2509402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3152061 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3152061 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0