| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600012138.mm.1 |
NM_198626.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012138.mm.1 |
NM_198626.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012138.mm.1 |
TC1600001471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012143.mm.1 |
NM_198626.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012143.mm.1 |
NM_198626.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012143.mm.1 |
TC1600001471.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012142.mm.1 |
NM_198626.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012142.mm.1 |
NM_198626.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012142.mm.1 |
TC1600001471.mm.1 |
MTA TC |