Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02394381_g1
          See other SULT2A2 GE Assays ›
          Gene Symbol:
          Sult2a2
          Gene Name:
          sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 2
          Gene Aliases:
          C730007P19Rik, ST, ST2A2, Sth2, mSTa2
          Chromosome Location:
          Chr.7: 13733506 - 13779637 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02394381_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sult2a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009286.2 NP_033312.2 2 272 154
          GenBank mRNA AK149424.1 - 2 273 154
          BC125365.1 - 245 40129880 154
          BC125367.1 - 245 40129882 154
          L27121.1 - 245 154

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sult2a2

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          biological_process
          steroid metabolic process
          molecular_function
          sulfotransferase activity
          transferase activity
          bile-salt sulfotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952512 BC125365.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952512 AK149424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952512 BC125367.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952512 L27121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952512 NM_009286.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172532 BC125365.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172532 BC125367.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172532 L27121.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111136 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105126 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952512 BC125365.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952512 AK149424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952512 BC125367.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952512 L27121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952512 NM_009286.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281568 BC125365.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281568 BC125367.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281568 L27121.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281568 BC147210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281568 NM_001081325.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06127044 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01480556 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105126 - -
          Whole Mouse Genome G4122A A_52_P430755 AI785533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P430755 NM_001184980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281568 BC125365.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281568 BC125367.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281568 L27121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281568 BC147210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281568 NM_001081325.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124633 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06127044 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105126 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01480556 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560114 - -
          Affymetrix HT MG-430 PM Array Plate 1419528_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560111 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550163 - -
          Mouse Genome 430 2.0 Array 1419528_at - -
          Mouse Genome 430 2.0 Array 1419528_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa11880_at - -
          Mouse Expression Set 430 1419528_at - -
          Mouse Expression Set 430 1419528_PM_at - -
          Murine Genome U74v2 Set 104484_at - -
          Murine Genome U74v2 Set 104484_PM_at - -
          Mouse Genome 430A 2.0 Array 1419528_PM_at - -
          Mouse Genome 430A 2.0 Array 1419528_at - -
          Affymetrix HT Mouse Genome 430A Array 1419528_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419528_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 619230 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2650397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002650397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242564 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242564 - -
          mouse-6 v1.1 (Illumina) scl020865.8_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 2650397 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020865.8_45-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2650397 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242564 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline