| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969530 |
BE981167.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969530 |
NM_011980.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
120074 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
799245 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726164 |
AK045344.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726164 |
NM_172868.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521373 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
102617 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155514 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969530 |
BE981167.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969530 |
NM_011980.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435879 |
BC127039.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435879 |
AJ224763.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435879 |
BE981167.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435879 |
AK047696.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435879 |
BC127038.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030191 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030208 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030192 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030207 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030201 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030204 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030205 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030202 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030199 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030194 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030197 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25030203 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P155514 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06153806 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P5803 |
BC127039.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P5803 |
AK047696.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P5803 |
AJ224763.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P5803 |
BC127038.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P5803 |
NM_011980.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435879 |
BC127039.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435879 |
AJ224763.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435879 |
BE981167.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435879 |
AK047696.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435879 |
BC127038.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P698754 |
AK037311.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104127 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P155514 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06456601 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06456595 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06456596 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030134 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030119 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030143 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030116 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030197 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030124 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030194 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030148 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030187 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030137 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030132 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030115 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030186 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030170 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030177 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030127 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030131 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030156 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030160 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030136 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030159 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030188 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030152 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030150 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030157 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030113 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030178 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030140 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030174 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030125 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030129 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030164 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030189 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030162 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030117 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25030145 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030191 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030123 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06347265 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06347269 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05902157 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030186 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06654763 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030162 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030178 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030189 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030192 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030207 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030113 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030202 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06347272 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030176 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030163 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06164908 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06151116 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030150 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030182 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030115 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030166 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030121 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030160 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030156 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06383065 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030197 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030119 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030174 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030180 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06139982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030117 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030181 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06276754 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030112 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06373102 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030184 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030201 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030204 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06492051 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030188 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030199 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06492054 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030116 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030179 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030187 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030205 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06383066 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06549101 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030159 |
- |
- |