Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525035_s1
          See other DEFB5 GE Assays ›
          Gene Symbol:
          Defb5
          Gene Name:
          defensin beta 5
          Gene Aliases:
          BD-5
          Chromosome Location:
          Chr.8: 19247592 - 19250828 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525035_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030734.2 NP_109659.2 2 137 82
          GenBank mRNA AF318068.1 - 137 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defb5

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          biological_process
          defense response to bacterium
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957669 NM_030734.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281168 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744125 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 179246 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957669 NM_030734.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25702595 - -
          Whole Mouse Genome G4122A A_51_P219017 NM_030734.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219017 AF318068.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07203920 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07203921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25702655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07326579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07281499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06876414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06694455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06720236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07203921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07009115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07326265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25702630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06983777 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10570613 - -
          Affymetrix HT MG-430 PM Array Plate 1421808_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10570612 - -
          Mouse Genome 430 2.0 Array 1421808_at - -
          Mouse Genome 430 2.0 Array 1421808_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81007_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81007_PM_at - -
          Mouse Expression Set 430 1421808_PM_at - -
          Mouse Expression Set 430 1421808_at - -
          Mouse Genome 430A 2.0 Array 1421808_PM_at - -
          Mouse Genome 430A 2.0 Array 1421808_at - -
          Affymetrix HT Mouse Genome 430A Array 1421808_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421808_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 401839 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3850047 - -
          Illumina mouse-6 v1.0 expression beadchip 3850047 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232541 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232541 - -
          mouse-6 v1.1 (Illumina) scl34017.2.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850047 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34017.2.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232541 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline