Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525218_s1
          See other CREBZF GE Assays ›
          Gene Symbol:
          Crebzf
          Gene Name:
          CREB/ATF bZIP transcription factor
          Gene Aliases:
          1110034C16Rik, 6330417B10Rik, LAZip, LAZipII, SMILE, Zf
          Chromosome Location:
          Chr.7: 90442735 - 90448043 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525218_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crebzf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145151.3 NP_660133.2 1 1577 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crebzf

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          response to virus
          negative regulation of transcription, DNA-templated
          regulation of sequence-specific DNA binding transcription factor activity
          transcription factor activity, sequence-specific DNA binding
          protein binding
          identical protein binding
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700011087.mm.1 TC0700001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011087.mm.1 NM_145151.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011087.mm.1 NR_073436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011093.mm.1 TC0700001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011093.mm.1 NM_145151.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011090.mm.1 TC0700001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011090.mm.1 NM_145151.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011091.mm.1 TC0700001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011091.mm.1 NM_145151.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011088.mm.1 TC0700001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011088.mm.1 NM_145151.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011088.mm.1 NR_073436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011088.mm.1 NR_073437.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011088.mm.1 NR_073438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011095.mm.1 TC0700001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011095.mm.1 NM_145151.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011094.mm.1 TC0700001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011094.mm.1 NM_145151.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline