Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525317_s1
          See other OLFR1356 GE Assays ›
          Gene Symbol:
          Olfr1356
          Gene Name:
          olfactory receptor 1356
          Gene Aliases:
          MOR142-1, MOR142-2_p
          Chromosome Location:
          Chr.10: 78846951 - 78847913 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525317_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1356
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146308.2 NP_666420.2 1 551 160

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1356

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363960 NM_146308.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363960 NM_146308.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128703 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363960 NM_146308.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363960 NM_146308.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363960 NM_146308.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363960 NM_146308.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275753 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128703 - -
          Whole Mouse Genome G4122A A_51_P363960 NM_146308.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363960 NM_146308.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101671 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128703 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08755011 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08723307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08759700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08723308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08723317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08570107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275762 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370540 - -
          Affymetrix Mouse Gene 1.1 ST Array 10370538 - -
          Affymetrix Mouse Clariom S Array PSR1000017944.mm.1 NM_146308.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017944.mm.1 NM_146308.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017944.mm.1 TC1000002462.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 887702 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6760609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257091 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257091 - -
          mouse-6 v1.1 (Illumina) scl37764.1.11_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 6760609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37764.1.11_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760609 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2715704 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2715704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline