Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525513_s1
          See other F630003A18RIK GE Assays ›
          Gene Symbol:
          F630003A18Rik
          Gene Name:
          RIKEN cDNA F630003A18 gene
          Gene Aliases:
          Cd200r5
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525513_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F630003A18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177010.3 NP_795984.1 7 1152 92
          GenBank mRNA AK089193.1 - 7 1153 92
          AK171165.1 - 1150 92
          BC147203.1 - 6 991 9056268 92
          BC147204.1 - 6 991 9056267 92

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene F630003A18Rik

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 AK171165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 BC147203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 BC147204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 NM_177010.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 AK089168.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 AK171165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 BC147203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 BC147204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 NM_177010.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P215638 AK089168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 AK171165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 BC147203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 BC147204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 NM_177010.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 AK089168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 AK171165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 BC147203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 BC147204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 NM_177010.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P215638 AK089168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 AK171165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 BC147203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 BC147204.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 NM_177010.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 AK089168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 AK171165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 BC147203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 BC147204.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 NM_177010.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215638 AK089168.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10435925 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608644 - -
          Affymetrix Mouse Gene 1.1 ST Array 10435907 - -
          Affymetrix Mouse Gene 1.1 ST Array 10435920 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 459822 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470270 - -
          Illumina mouse-6 v1.0 expression beadchip 2470270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737226 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737226 - -
          mouse-6 v1.1 (Illumina) scl00319823.1_85-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470270 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00319823.1_85-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470270 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2831116 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2831116 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0