Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525632_s1
          See other RRAGA GE Assays ›
          Gene Symbol:
          Rraga
          Gene Name:
          Ras-related GTP binding A
          Gene Aliases:
          1300010C19Rik, AI255374, FIP-1, RAGA
          Chromosome Location:
          Chr.4: 86575673 - 86577285 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525632_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rraga
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178376.3 NP_848463.1 1 1377 135
          GenBank mRNA AK004955.1 - 1 1339 135
          AK144591.1 - 1 1306 135
          BC037615.1 - 1 1345 135
          BC048245.1 - 1 1367 6311881 135

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rraga

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          cell death
          cellular response to starvation
          regulation of autophagy
          modulation by virus of host morphology or physiology
          positive regulation of TOR signaling
          cellular protein localization
          regulation of cytolysis
          positive regulation of cytolysis
          cellular response to amino acid stimulus
          nucleotide binding
          GTPase activity
          GTP binding
          protein homodimerization activity
          protein heterodimerization activity
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080555 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080558 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080559 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080556 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 BC048245.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 NM_178376.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 AK004955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 AK144591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 BC037615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 BC048245.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 NM_178376.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 AK004955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 AK144591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226417 BC037615.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68277 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125009 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 BC048245.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 NM_178376.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 AK004955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 AK144591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 BC037615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 BC048245.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 NM_178376.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 AK004955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 AK144591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226417 BC037615.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 BC048245.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 NM_178376.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 AK004955.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 AK144591.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 BC037615.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 BC048245.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 NM_178376.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 AK004955.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 AK144591.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226417 BC037615.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080556 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125009 - -
          Whole Mouse Genome G4122A A_51_P226417 BC048245.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 NM_178376.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 AK004955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 AK144591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 BC037615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 BC048245.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 NM_178376.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 AK004955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 AK144591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226417 BC037615.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112475 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125009 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03610962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03610964 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03610959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03610949 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080559 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080558 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23080555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03635341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03988451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03635340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03681257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03965253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03640047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080568 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505748 - -
          Affymetrix HT MG-430 PM Array Plate 1428905_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10505747 - -
          Mouse Genome 430 2.0 Array 1428905_at - -
          Mouse Genome 430 2.0 Array 1428905_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68441_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68441_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400005893.mm.1 NM_178376.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005893.mm.1 NM_178376.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005893.mm.1 TC0400000867.mm.1 MTA TC
          Mouse Expression Set 430 1428905_PM_at - -
          Mouse Expression Set 430 1428905_at - -
          Murine Genome U74v2 Set 94257_at - -
          Murine Genome U74v2 Set 94257_PM_at - -
          Mouse Genome 430A 2.0 Array 1428905_at - -
          Mouse Genome 430A 2.0 Array 1428905_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428905_at - -
          Affymetrix HT Mouse Genome 430A Array 1428905_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 341113 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6400603 - -
          Illumina mouse-6 v1.0 expression beadchip 6400603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006400603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2757931 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2757931 - -
          mouse-6 v1.1 (Illumina) scl25301.1.41_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6400603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25301.1.41_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2757931 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2757931 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25301.1.41_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0