Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525932_s1
          See other TCSTV1 GE Assays ›
          Gene Symbol:
          Tcstv1
          Gene Name:
          2-cell-stage, variable group, member 1
          Gene Aliases:
          Lo1
          Chromosome Location:
          Chr.13: 119893387 - 119894785 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525932_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcstv1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018756.3 NP_061226.1 2 758 145
          GenBank mRNA AF067057.1 - 2 740 145
          AF067059.1 - 2 758 145
          BC119476.2 - 1 1242 40049850 145

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcstv1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481768 AF067057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481768 AF067059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481768 BC119476.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481768 NM_018756.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481768 AF067057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481768 AF067059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481768 BC119476.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P481768 NM_018756.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481768 AF067057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481768 AF067059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481768 BC119476.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481768 NM_018756.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481768 AF067057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481768 AF067059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481768 BC119476.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P481768 NM_018756.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481768 AF067057.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481768 AF067059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481768 BC119476.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481768 NM_018756.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481768 AF067057.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481768 AF067059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481768 BC119476.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481768 NM_018756.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481777 AF067057.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481777 AF067059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481777 BC119476.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P481777 NM_018756.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P66425 AF067057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P66425 AF067059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P66425 BC119476.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P66425 NM_018756.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481768 AF067057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481768 AF067059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481768 BC119476.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481768 NM_018756.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481768 AF067057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481768 AF067059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481768 BC119476.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481768 NM_018756.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481777 AF067057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481777 AF067059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481777 BC119476.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P481777 NM_018756.2 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598150 - -
          Affymetrix HT MG-430 PM Array Plate 1420729_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608670 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598146 - -
          Mouse Genome 430 2.0 Array 1420729_at - -
          Mouse Genome 430 2.0 Array 1420729_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300018613.mm.1 TC1300002751.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018614.mm.1 NM_018756.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018614.mm.1 TC1300002751.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018611.mm.1 NM_018756.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018611.mm.1 TC1300002751.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018612.mm.1 NM_018756.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018612.mm.1 TC1300002751.mm.1 MTA TC
          Mouse Expression Set 430 1420729_at - -
          Mouse Expression Set 430 1420729_PM_at - -
          Murine Genome U74v2 Set 94727_f_at - -
          Murine Genome U74v2 Set 94727_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1420729_at - -
          Mouse Genome 430A 2.0 Array 1420729_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420729_at - -
          Affymetrix HT Mouse Genome 430A Array 1420729_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 801958 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 730465 - -
          Illumina mouse-6 v1.0 expression beadchip 730465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730465 - -
          mouse-6 v1.1 (Illumina) scl054382.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 730465 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl054382.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730465 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2946986 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241056 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2946986 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0