Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525960_s1
          See other IFNA7 GE Assays ›
          Gene Symbol:
          Ifna7
          Gene Name:
          interferon alpha 7
          Gene Aliases:
          Ifa7
          Chromosome Location:
          Chr.4: 88816225 - 88816800 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525960_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifna7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008334.3 NP_032360.2 1 237 134
          GenBank mRNA BC130231.1 - 1 238 40141930 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000904.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifna7

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          cytokine activity
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030186 BC130231.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030186 NM_008334.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030186 BC130231.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030186 NM_008334.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092346 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03620075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03370592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03747043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092315 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505880 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505879 - -
          Affymetrix Mouse Gene 1.1 ST Array 10514289 - -
          Mouse Genome 430 2.0 Array 1422403_PM_at - -
          Mouse Genome 430 2.0 Array 1422403_at - -
          Mouse Genome 430 2.0 Array 1422404_x_at - -
          Mouse Genome 430 2.0 Array 1422404_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR0400006096.mm.1 NM_008334.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006096.mm.1 NM_008334.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006096.mm.1 TC0400000904.mm.1 MTA TC
          Mouse Expression Set 430 1422404_x_at - -
          Mouse Expression Set 430 1422404_PM_x_at - -
          Mouse Expression Set 430 1450593_at - -
          Mouse Expression Set 430 1422403_at - -
          Mouse Expression Set 430 1422403_PM_at - -
          Mouse Expression Set 430 1450593_PM_at - -
          Murine Genome U74v2 Set 102149_PM_f_at - -
          Murine Genome U74v2 Set 102149_f_at - -
          Murine Genome U74v2 Set 101619_at - -
          Murine Genome U74v2 Set 96419_PM_f_at - -
          Murine Genome U74v2 Set 101619_PM_at - -
          Murine Genome U74v2 Set 96419_f_at - -
          Mouse Genome 430A 2.0 Array 1422403_PM_at - -
          Mouse Genome 430A 2.0 Array 1422404_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422403_at - -
          Mouse Genome 430A 2.0 Array 1450593_at - -
          Mouse Genome 430A 2.0 Array 1422404_x_at - -
          Mouse Genome 430A 2.0 Array 1450593_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2510239 - -
          Illumina mouse-6 v1.0 expression beadchip 106040181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002510239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658633 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658633 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235789 - -
          mouse-6 v1.1 (Illumina) GI_45598383-S - -
          mouse-6 v1.1 (Illumina) scl015970.1_178-S - -
          Illumina Sentrix mouse-6.v1 bead array 106040181 - -
          Illumina Sentrix mouse-6.v1 bead array 2510239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015970.1_178-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106040181 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658633 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658633 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0