Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02526089_u1
          See other KLRA19 GE Assays ›
          Gene Symbol:
          Klra19
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 19
          Gene Aliases:
          Ly49s
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02526089_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053154.2 NP_444384.1 4 329 124
          GenBank mRNA AF288378.1 - 4 329 124
          BC116822.1 - 4 288 40126141 124
          BC116824.1 - 4 282 40126143 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003253.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra19

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P216488 NM_053154.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P216488 AF288378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P216488 BC116822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P216488 BC116824.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P216488 NM_053154.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P216488 AF288378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P216488 BC116822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P216488 BC116824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P216488 NM_053154.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P216488 AF288378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P216488 BC116822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P216488 BC116824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P216488 NM_053154.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P216488 AF288378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P216488 BC116822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P216488 BC116824.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P216488 NM_053154.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P216488 AF288378.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P216488 BC116822.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P216488 BC116824.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P216488 NM_053154.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P216488 AF288378.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P216488 BC116822.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P216488 BC116824.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846251 - -
          Whole Mouse Genome G4122A A_52_P216488 NM_053154.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P216488 AF288378.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P216488 BC116822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P216488 BC116824.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P216488 NM_053154.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P216488 AF288378.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P216488 BC116822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P216488 BC116824.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05604856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05321444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05139424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05321444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05550144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05550141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05604858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548503 - -
          Affymetrix HT MG-430 PM Array Plate 1426140_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608686 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548513 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548525 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548497 - -
          Mouse Genome 430 2.0 Array 1426140_x_at - -
          Mouse Genome 430 2.0 Array 1426140_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR0600025531.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025531.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025531.mm.1 NM_008464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025532.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025532.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025532.mm.1 NM_008464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025537.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025537.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025537.mm.1 NM_008464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025533.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025533.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025533.mm.1 NM_008464.2 Refseq
          Mouse Expression Set 430 1426140_x_at - -
          Mouse Expression Set 430 1426140_PM_x_at - -
          Murine Genome U74v2 Set 100674_PM_f_at - -
          Murine Genome U74v2 Set 100674_f_at - -
          Mouse Genome 430A 2.0 Array 1426140_x_at - -
          Mouse Genome 430A 2.0 Array 1426140_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426140_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426140_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 315669 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2981486 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2981486 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0