Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02526725_s1
          See other OMG GE Assays ›
          Gene Symbol:
          Omg
          Gene Name:
          oligodendrocyte myelin glycoprotein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 79500982 - 79504082 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02526725_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Omg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019409.2 NP_062282.2 2 1610 128
          GenBank mRNA AK039141.1 - 1 977 128
          AK136910.1 - 2 1613 128
          BC024757.1 - 2 1503 5369312 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Omg

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          central nervous system myelination
          neuron projection regeneration
          regulation of collateral sprouting of intact axon in response to injury
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105472 BC024757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105472 NM_019409.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105472 AK039141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105472 AK136910.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123882 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105472 BC024757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105472 NM_019409.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105472 AK039141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105472 AK136910.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P255832 BC024757.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P255832 NM_019409.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P255832 AK039141.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P255832 AK136910.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949239 - -
          Whole Mouse Genome G4122A A_51_P255832 BC024757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P255832 NM_019409.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P255832 AK039141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P255832 AK136910.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136437 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09621424 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09621422 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09621426 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09537898 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09351061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09351060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09414428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09001808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09621423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09414426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09537897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388955 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388957 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388956 - -
          Affymetrix HT MG-430 PM Array Plate 1418212_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10388954 - -
          Mouse Genome 430 2.0 Array 1418212_PM_at - -
          Mouse Genome 430 2.0 Array 1418212_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18377_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18377_at - -
          Affymetrix Mouse Clariom S Array PSR1100030592.mm.1 NM_019409.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030592.mm.1 NM_019409.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030592.mm.1 TC1100003325.mm.1 MTA TC
          Mouse Expression Set 430 1418212_PM_at - -
          Mouse Expression Set 430 1418212_at - -
          Mouse Genome 430A 2.0 Array 1418212_at - -
          Mouse Genome 430A 2.0 Array 1418212_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418212_at - -
          Affymetrix HT Mouse Genome 430A Array 1418212_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 681282 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6760066 - -
          Illumina mouse-6 v1.0 expression beadchip 102340324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2603450 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2603450 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248920 - -
          mouse-6 v1.1 (Illumina) scl0018377.2_15-S - -
          mouse-6 v1.1 (Illumina) scl39866.2_254-S - -
          Illumina Sentrix mouse-6.v1 bead array 6760066 - -
          Illumina Sentrix mouse-6.v1 bead array 102340324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0018377.2_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102340324 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2603450 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248920 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2603450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0018377.2_15-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline