| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10474813 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10474810 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10474811 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10474812 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1418871_PM_a_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10474809 |
- |
- |
| Mouse Genome 430 2.0 Array |
1418871_a_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1418871_PM_a_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0200012940.mm.1 |
NM_001145644.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012940.mm.1 |
NM_001145644.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012940.mm.1 |
TC0200001746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012938.mm.1 |
NM_001145644.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012938.mm.1 |
NM_001145644.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012938.mm.1 |
TC0200001746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012949.mm.1 |
NM_001145644.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012949.mm.1 |
NM_001145644.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012949.mm.1 |
TC0200001746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012950.mm.1 |
NM_001145644.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012950.mm.1 |
NM_001145644.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012950.mm.1 |
TC0200001746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012952.mm.1 |
NM_001145644.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012952.mm.1 |
NM_001145644.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012952.mm.1 |
TC0200001746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012954.mm.1 |
NM_001145644.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012954.mm.1 |
NM_001145644.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012954.mm.1 |
TC0200001746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200012953.mm.1 |
NM_001145644.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012953.mm.1 |
NM_001145644.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200012953.mm.1 |
TC0200001746.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1418871_PM_a_at |
- |
- |
| Mouse Expression Set 430 |
1418871_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1418871_PM_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1418871_a_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1418871_a_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1418871_PM_a_at |
- |
- |