Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02527285_s1
          See other TRIM59 GE Assays ›
          Gene Symbol:
          Trim59
          Gene Name:
          tripartite motif-containing 59
          Gene Aliases:
          2310035M22Rik, 2700022F13Rik, Mrf1, TSBF1
          Chromosome Location:
          Chr.3: 69035292 - 69046404 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02527285_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trim59
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025863.3 NP_080139.3 3 1941 153
          GenBank mRNA AK009637.1 - 1 1014 153
          BC006700.1 - 3 1859 3501285 153
          BC025430.1 - 3 1874 4017983 153
          BE847098.1 - 1 292 153

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trim59

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          negative regulation of viral entry into host cell
          protein binding
          zinc ion binding
          metal ion binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218906 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218903 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218902 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218908 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218909 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218901 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218904 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218907 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030938 BC025430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030938 BC006700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030938 NM_025863.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030938 AK009637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105152 BC025430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105152 BC006700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105152 NM_025863.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105152 AK009637.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9894 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168574 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105152 BC025430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105152 BC006700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105152 NM_025863.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105152 AK009637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030938 BC025430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030938 BC006700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030938 NM_025863.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030938 AK009637.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22218915 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04940894 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168574 - -
          Whole Mouse Genome G4122A A_52_P512561 BC025430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P512561 BC055347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P512561 CD541510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P512561 AK016642.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P512561 BC006700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275808 BC025430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275808 BC006700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275808 NM_025863.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275808 AK009637.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129765 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168574 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02319272 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02849747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02849746 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03061846 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03061847 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03061853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218906 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218902 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218907 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218908 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218909 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218901 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218904 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22218903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22218947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02294048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03175672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02863407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02883501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02831601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02393627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02319272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02849748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02884888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02320978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03048016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02774559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02774542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02666910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22218892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10498622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498621 - -
          Affymetrix HT MG-430 PM Array Plate 1431430_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1416118_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10498620 - -
          Mouse Genome 430 2.0 Array 1443354_at - -
          Mouse Genome 430 2.0 Array 1431430_s_at - -
          Mouse Genome 430 2.0 Array 1416118_PM_at - -
          Mouse Genome 430 2.0 Array 1431430_PM_s_at - -
          Mouse Genome 430 2.0 Array 1416118_at - -
          Mouse Genome 430 2.0 Array 1443354_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66949_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66949_at - -
          Affymetrix Mouse Clariom S Array PSR0300016050.mm.1 NM_025863.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016050.mm.1 NM_025863.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016050.mm.1 TC0300002126.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016051.mm.1 NM_025863.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016051.mm.1 NM_025863.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016051.mm.1 TC0300002126.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016055.mm.1 NM_025863.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016055.mm.1 NM_025863.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016055.mm.1 TC0300002126.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016060.mm.1 NM_025863.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016060.mm.1 NM_025863.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016060.mm.1 TC0300002126.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016062.mm.1 NM_025863.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016062.mm.1 NM_025863.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016062.mm.1 TC0300002126.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016049.mm.1 NM_025863.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016049.mm.1 NM_025863.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016049.mm.1 TC0300002126.mm.1 MTA TC
          Mouse Expression Set 430 1416118_PM_at - -
          Mouse Expression Set 430 1416118_at - -
          Mouse Expression Set 430 1431430_PM_s_at - -
          Mouse Expression Set 430 1431430_s_at - -
          Murine Genome U74v2 Set 99149_PM_at - -
          Murine Genome U74v2 Set 99149_at - -
          Mouse Genome 430A 2.0 Array 1416118_at - -
          Mouse Genome 430A 2.0 Array 1416118_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 360918 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5570072 - -
          Illumina mouse-6 v1.0 expression beadchip 5570072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570072 - -
          mouse-6 v1.1 (Illumina) scl22076.2_386-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22076.2_386-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2993661 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2993661 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2515081 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline