Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02527500_g1
          See other KRTAP5-2 GE Assays ›
          Gene Symbol:
          Krtap5-2
          Gene Name:
          keratin associated protein 5-2
          Gene Aliases:
          4833428E21Rik
          Chromosome Location:
          Chr.7: 142174532 - 142176005 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02527500_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krtap5-2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027844.4 NP_082120.3 1 1368 86
          GenBank mRNA AK014785.1 - 2 835 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004557.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krtap5-2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154987 AK014785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985298 BC148222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985298 AK014785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985298 NM_027844.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154987 AK014785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985298 BC148222.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985298 AK014785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985298 NM_027844.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593295 - -
          Whole Mouse Genome G4122A A_51_P355981 AK014785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355981 NM_027844.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06337154 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06337157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06206855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06337157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06270757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06494329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593295 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569295 - -
          Affymetrix HT MG-430 PM Array Plate 1420452_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569296 - -
          Affymetrix Mouse Gene 1.1 ST Array 10569291 - -
          Affymetrix Mouse Gene 1.1 ST Array 10569288 - -
          Mouse Genome 430 2.0 Array 1420452_PM_at - -
          Mouse Genome 430 2.0 Array 1420452_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71623_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71623_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700038429.mm.1 NM_027844.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038429.mm.1 NM_027844.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038429.mm.1 TC0700004557.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038427.mm.1 NM_027844.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038427.mm.1 NM_027844.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038427.mm.1 TC0700004557.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038428.mm.1 NM_027844.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038428.mm.1 NM_027844.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038428.mm.1 TC0700004557.mm.1 MTA TC
          Mouse Expression Set 430 1420452_at - -
          Mouse Expression Set 430 1420452_PM_at - -
          Mouse Genome 430A 2.0 Array 1420452_PM_at - -
          Mouse Genome 430A 2.0 Array 1420452_at - -
          Affymetrix HT Mouse Genome 430A Array 1420452_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420452_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 542943 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3850286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694941 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694941 - -
          mouse-6 v1.1 (Illumina) scl071623.1_73-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl071623.1_73-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2800530 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2800530 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline