Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02527814_s1
          See other CML2 GE Assays ›
          Gene Symbol:
          Cml2
          Gene Name:
          camello-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 85865422 - 85869137 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02527814_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cml2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053096.3 NP_444326.2 3 1458 79
          GenBank mRNA BC039773.1 - 3 1478 4193874 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cml2

          Panther Classification:

          Molecular Function -

          acetyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          gastrulation with mouth forming second
          negative regulation of cell adhesion
          multicellular organism development
          gastrulation
          dihydrolipoamide branched chain acyltransferase activity
          sterol O-acyltransferase activity
          N-acetyltransferase activity
          O-acyltransferase activity
          palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
          carnitine O-acyltransferase activity
          acetyltransferase activity
          C-acyltransferase activity
          palmitoyltransferase activity
          N-acyltransferase activity
          acylglycerol O-acyltransferase activity
          serine O-acyltransferase activity
          O-acetyltransferase activity
          O-octanoyltransferase activity
          octanoyltransferase activity
          O-palmitoyltransferase activity
          S-acyltransferase activity
          S-acetyltransferase activity
          S-malonyltransferase activity
          malonyltransferase activity
          C-palmitoyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          succinyltransferase activity
          N-succinyltransferase activity
          O-succinyltransferase activity
          S-succinyltransferase activity
          sinapoyltransferase activity
          O-sinapoyltransferase activity
          peptidyl-lysine N6-myristoyltransferase activity
          peptidyl-lysine N6-palmitoyltransferase activity
          benzoyl acetate-CoA thiolase activity
          3-hydroxybutyryl-CoA thiolase activity
          3-ketopimelyl-CoA thiolase activity
          N-palmitoyltransferase activity
          myristoyltransferase activity
          acyl-CoA N-acyltransferase activity
          protein-cysteine S-myristoyltransferase activity
          protein-cysteine S-acyltransferase activity
          dihydrolipoamide S-acyltransferase activity
          glucosaminyl-phosphotidylinositol O-acyltransferase activity
          ergosterol O-acyltransferase activity
          lanosterol O-acyltransferase activity
          naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
          2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
          2-methylhexanoyl-CoA C-acetyltransferase activity
          butyryl-CoA 2-C-propionyltransferase activity
          2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
          L-2-aminoadipate N-acetyltransferase activity
          UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
          keto acid formate lyase activity
          Ras palmitoyltransferase activity
          azetidine-2-carboxylic acid acetyltransferase activity
          peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
          acetyl-CoA:L-lysine N6-acetyltransferase

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P240712 BC039773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P240712 NM_053096.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P240712 BC039773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P240712 NM_053096.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92722 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111029 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P240712 BC039773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P240712 NM_053096.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P240712 BC039773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P240712 NM_053096.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P206683 BC039773.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P206683 NM_053096.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P206683 AF163315.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P240712 BC039773.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P240712 NM_053096.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P240712 BC039773.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P240712 NM_053096.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206683 BC039773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206683 NM_053096.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206683 AF163315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P240712 BC039773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P240712 NM_053096.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P240712 BC039773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P240712 NM_053096.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127303 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05623394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24606874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24606876 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545885 - -
          Affymetrix HT MG-430 PM Array Plate 1421108_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455232_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1421109_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545881 - -
          Mouse Genome 430 2.0 Array 1421108_at - -
          Mouse Genome 430 2.0 Array 1455232_at - -
          Mouse Genome 430 2.0 Array 1421109_at - -
          Mouse Genome 430 2.0 Array 1421109_PM_at - -
          Mouse Genome 430 2.0 Array 1421108_PM_at - -
          Mouse Genome 430 2.0 Array 1455232_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93673_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93673_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600020369.mm.1 TC0600002624.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020369.mm.1 NM_053096.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020369.mm.1 NM_053096.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020370.mm.1 TC0600002624.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020370.mm.1 NM_053096.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020370.mm.1 NM_053096.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020371.mm.1 TC0600002624.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020371.mm.1 NM_053096.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020371.mm.1 NM_053096.3 Refseq
          Mouse Expression Set 430 1421108_PM_at - -
          Mouse Expression Set 430 1455232_at - -
          Mouse Expression Set 430 1421109_at - -
          Mouse Expression Set 430 1455232_PM_at - -
          Mouse Expression Set 430 1421108_at - -
          Mouse Expression Set 430 1421109_PM_at - -
          Murine Genome U74v2 Set 130761_at - -
          Murine Genome U74v2 Set 130761_PM_at - -
          Mouse Genome 430A 2.0 Array 1421108_PM_at - -
          Mouse Genome 430A 2.0 Array 1421109_PM_at - -
          Mouse Genome 430A 2.0 Array 1421109_at - -
          Mouse Genome 430A 2.0 Array 1455232_PM_at - -
          Mouse Genome 430A 2.0 Array 1455232_at - -
          Mouse Genome 430A 2.0 Array 1421108_at - -
          Affymetrix HT Mouse Genome 430A Array 1421108_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421109_at - -
          Affymetrix HT Mouse Genome 430A Array 1421108_at - -
          Affymetrix HT Mouse Genome 430A Array 1455232_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421109_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455232_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 522898 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550315 - -
          Illumina mouse-6 v1.0 expression beadchip 1170504 - -
          Illumina mouse-6 v1.0 expression beadchip 2340458 - -
          Illumina mouse-6 v1.0 expression beadchip 5550315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170504 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550315 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2734410 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248507 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215139 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734410 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248507 - -
          mouse-6 v1.1 (Illumina) scl28761.2_42-S - -
          mouse-6 v1.1 (Illumina) scl093673.1_29-S - -
          mouse-6 v1.1 (Illumina) scl0093673.1_94-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550315 - -
          Illumina Sentrix mouse-6.v1 bead array 2340458 - -
          Illumina Sentrix mouse-6.v1 bead array 1170504 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0093673.1_94-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl093673.1_29-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28761.2_42-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170504 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340458 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248507 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2734410 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215139 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2734410 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248507 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0