Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02528568_g1
          See other 9930111J21RIK2 GE Assays ›
          Gene Symbol:
          9930111J21Rik2
          Gene Name:
          RIKEN cDNA 9930111J21 gene 2
          Gene Aliases:
          9930111J21Rik, Irgb5, OTTMUSG00000005540
          Chromosome Location:
          Chr.11: 49037660 - 49051242 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02528568_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9930111J21Rik2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173434.1 NP_775610.1 3 1648 68
          GenBank mRNA AK037088.1 - 3 1648 68
          BC119805.1 - 2 1552 40107440 68
          BC122882.1 - 2 1557 40107441 68

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 9930111J21Rik2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61697 BC119805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61697 BC122882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61697 AK037088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61697 NM_173434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61697 BC119805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61697 BC122882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61697 AK037088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P61697 NM_173434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61697 BC119805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61697 BC122882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61697 AK037088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61697 NM_173434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61697 BC119805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61697 BC122882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61697 AK037088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P61697 NM_173434.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07334600 - -
          Whole Mouse Genome G4122A A_52_P61697 BC119805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61697 BC122882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61697 AK037088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61697 NM_173434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61697 BC119805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61697 BC122882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61697 AK037088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P61697 NM_173434.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07334600 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385532 - -
          Affymetrix Mouse Gene 1.1 ST Array 10385526 - -
          Affymetrix Mouse Gene 1.1 ST Array 10385513 - -
          Affymetrix Mouse Clariom S Array PSR1100024214.mm.1 NM_173434.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024214.mm.1 NM_173434.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024214.mm.1 TC1100002648.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024213.mm.1 NM_173434.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024213.mm.1 NM_173434.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024213.mm.1 TC1100002648.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024199.mm.1 TC1100004292.mm.1 MTA TC
          Mouse Expression Set 430 1425728_PM_at - -
          Mouse Expression Set 430 1425728_at - -
          Mouse Genome 430A 2.0 Array 1425728_at - -
          Mouse Genome 430A 2.0 Array 1425728_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 865250 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4280056 - -
          Illumina mouse-6 v1.0 expression beadchip 1230193 - -
          Illumina mouse-6 v1.0 expression beadchip 4280056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249820 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258795 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249820 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258795 - -
          mouse-6 v1.1 (Illumina) scl0245240.1_49-S - -
          mouse-6 v1.1 (Illumina) scl00245240.2_95-S - -
          Illumina Sentrix mouse-6.v1 bead array 4280056 - -
          Illumina Sentrix mouse-6.v1 bead array 1230193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0245240.1_49-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00245240.2_95-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280056 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2685088 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249820 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2685088 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline