| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10587613 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10587614 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10587615 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10587612 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10587611 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10587610 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900008784.mm.1 |
NR_045835.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008784.mm.1 |
NR_045835.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008784.mm.1 |
TC0900001116.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900008782.mm.1 |
NR_045835.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008782.mm.1 |
NR_045835.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008782.mm.1 |
TC0900001116.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900008783.mm.1 |
NR_045835.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008783.mm.1 |
NR_045835.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008783.mm.1 |
TC0900001116.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900008785.mm.1 |
NR_045835.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008785.mm.1 |
NR_045835.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008785.mm.1 |
TC0900001116.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900008786.mm.1 |
NR_045835.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008786.mm.1 |
NR_045835.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008786.mm.1 |
TC0900001116.mm.1 |
MTA TC |