Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02529917_s1
          See other CALM5 GE Assays ›
          Gene Symbol:
          Calm5
          Gene Name:
          calmodulin 5
          Gene Aliases:
          AY314008, Scarf2
          Chromosome Location:
          Chr.13: 3854173 - 3854761 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02529917_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Calm5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008706.2 NP_001008706.1 2 452 103
          GenBank mRNA AY314008.1 - 357 103
          BC138691.1 - 1 491 8861713 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Calm5

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P329414 NM_001008706.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P329414 AY314008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P329414 BC138691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P329414 NM_001008706.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P329414 AY314008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P329414 BC138691.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25104 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117478 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145301 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P329414 NM_001008706.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P329414 AY314008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P329414 BC138691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P329414 NM_001008706.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P329414 AY314008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P329414 BC138691.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790147 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117478 - -
          Whole Mouse Genome G4122A A_52_P329414 NM_001008706.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P329414 AY314008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P329414 BC138691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P329414 NM_001008706.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P329414 AY314008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P329414 BC138691.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10874389 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10874408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10577281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10577277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10867753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790116 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403284 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403283 - -
          Affymetrix Mouse Clariom S Array PSR1300000101.mm.1 NM_001008706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000101.mm.1 NM_001008706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000101.mm.1 TC1300000014.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160926 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160926 - -
          mouse-6 v1.1 (Illumina) GI_56785417-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160926 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160926 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline