Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581132_s1
          See other OLFR1418 GE Assays ›
          Gene Symbol:
          Olfr1418
          Gene Name:
          olfactory receptor 1418
          Gene Aliases:
          MOR266-5
          Chromosome Location:
          Chr.19: 11854995 - 11855951 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02581132_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1418
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011524.1 NP_001011524.1 1 903 84
          GenBank mRNA BC150977.1 - 1 953 9087890 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001146.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1418

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103185 BC150977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103185 NM_001011524.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144399 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103185 BC150977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103185 NM_001011524.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958163 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144399 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144399 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14506967 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14260848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14506968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14248197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14506968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14298313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14223477 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466269 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466267 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466268 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466266 - -
          Affymetrix Mouse Clariom S Array PSR1900010750.mm.1 TC1900001146.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160297 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160297 - -
          mouse-6 v1.1 (Illumina) GI_58372129-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160297 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160297 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jvxmr:80/100.66.77.199:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0