Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581433_mH
          See other GM4788 GE Assays ›
          Gene Symbol:
          Gm4788
          Gene Name:
          predicted gene 4788
          Gene Aliases:
          Cfhrc, EG214403
          Chromosome Location:
          Chr.1: 139697919 - 139781239 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02581433_mH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm4788
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029977.3 NP_001025148.3 1 - 2 148 101
          NM_001160303.1 NP_001153775.1 1 - 2 148 101
          NM_001160304.1 NP_001153776.1 1 - 2 148 101
          GenBank mRNA AJ871401.1 - 1 - 2 77 101
          AJ871402.1 - 1 - 2 77 101
          BC141136.1 - 1 - 2 120 9055748 101
          BC145444.1 - 1 - 2 120 9054002 101
          M29009.1 - 1 - 2 145 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003190.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm4788

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017413 BC145444.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017413 M29009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017413 AJ871402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017413 BC141136.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017413 AJ871401.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441852 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502664 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623559 AK046955.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623559 NM_198659.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85018 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109497 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140243 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017413 BC145444.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017413 M29009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017413 AJ871402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017413 BC141136.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017413 AJ871401.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P512525 BC145444.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P512525 M29009.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P512525 AJ871402.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P512525 BC141136.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P512525 AJ871401.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04284745 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04284730 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00200921 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04284750 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140243 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04284752 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00200917 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109497 - -
          Whole Mouse Genome G4122A A_51_P512519 M29009.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00200921 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04284745 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140243 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04284752 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00378302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20663869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20663969 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10358305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358307 - -
          Affymetrix HT MG-430 PM Array Plate 1425999_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10358299 - -
          Mouse Genome 430 2.0 Array 1425999_at - -
          Mouse Genome 430 2.0 Array 1425999_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100025861.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025861.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025861.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025861.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025861.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025861.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025861.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025866.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025866.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025866.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025866.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025866.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025866.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025866.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025863.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025863.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025863.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025863.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025863.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025863.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025863.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025872.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025872.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025872.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025872.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025872.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025872.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025872.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025873.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025873.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025873.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025873.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025873.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025873.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025873.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025862.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025862.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025862.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025862.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025862.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025875.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025875.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025875.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025875.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025875.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025875.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025875.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025865.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025865.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025865.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025865.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025865.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025865.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025865.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025868.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025868.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025868.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025868.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025868.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025868.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025868.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025871.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025871.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025871.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025871.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025871.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025871.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025871.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025874.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025874.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025874.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025874.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025874.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025874.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025874.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025864.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025864.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025864.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025864.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025864.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025864.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025864.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025870.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025870.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025870.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025870.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025870.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025870.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025870.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025876.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025876.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025876.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025876.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025876.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025876.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025876.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025860.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025860.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025860.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025860.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025860.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025860.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025860.mm.1 TC0100003190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100025867.mm.1 NM_001029977.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025867.mm.1 NM_001029977.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025867.mm.1 NM_001160303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025867.mm.1 NM_001160303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025867.mm.1 NM_001160304.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025867.mm.1 NM_001160304.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100025867.mm.1 TC0100003190.mm.1 MTA TC
          Mouse Expression Set 430 1425999_at - -
          Mouse Expression Set 430 1425999_PM_at - -
          Murine Genome U74v2 Set 94743_PM_f_at - -
          Murine Genome U74v2 Set 94743_f_at - -
          Mouse Genome 430A 2.0 Array 1425999_at - -
          Mouse Genome 430A 2.0 Array 1425999_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425999_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425999_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162960 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162960 - -
          mouse-6 v1.1 (Illumina) GI_71533185-I - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162960 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162960 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline