| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10598140 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10598139 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1438073_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10598138 |
- |
- |
| Mouse Genome 430 2.0 Array |
1438073_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1438073_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000032.mm.1 |
NM_001030293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000032.mm.1 |
TCX_GL456233_random00000007.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000031.mm.1 |
NM_001030293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000031.mm.1 |
TCX_GL456233_random00000007.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000029.mm.1 |
NM_001030293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000029.mm.1 |
TCX_GL456233_random00000007.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000030.mm.1 |
NM_001030293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000030.mm.1 |
TCX_GL456233_random00000007.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000033.mm.1 |
NM_001030293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000033.mm.1 |
TCX_GL456233_random00000007.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000028.mm.1 |
NM_001030293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000028.mm.1 |
TCX_GL456233_random00000007.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1438073_at |
- |
- |
| Mouse Expression Set 430 |
1438073_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1438073_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1438073_PM_at |
- |
- |