Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581554_m1
          See other CSN3 GE Assays ›
          Gene Symbol:
          Csn3
          Gene Name:
          casein kappa
          Gene Aliases:
          AW208918, CSN10, Csnk
          Chromosome Location:
          Chr.5: 87925575 - 87932545 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02581554_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007786.4 NP_031812.2 4 - 5 637 99
          GenBank mRNA AK142673.1 - 4 - 5 638 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000802.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csn3

          Panther Classification:

          Molecular Function -

          storage protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lactation
          protein stabilization
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034496 AK142673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2034496 NM_007786.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129853 AK021350.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 BC049949.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 BC050269.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 M10114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 AK164788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 AK085710.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 BC049949.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 BC050269.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 M10114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 AK164788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P227275 AK085710.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326743 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846237 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134039 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034496 AK142673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2034496 NM_007786.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870334 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645933 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134039 - -
          Whole Mouse Genome G4122A A_51_P227280 M10114.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129429 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134039 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04683717 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04683716 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04683709 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04683721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04683707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04768898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04111669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04116878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04788448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04116885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04307977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04606518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04768897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04653974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04677875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04307978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870346 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10522901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522896 - -
          Affymetrix HT MG-430 PM Array Plate 1419735_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451851_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10522895 - -
          Mouse Genome 430 2.0 Array 1451851_a_at - -
          Mouse Genome 430 2.0 Array 1419735_PM_at - -
          Mouse Genome 430 2.0 Array 1419735_at - -
          Mouse Genome 430 2.0 Array 1451851_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12994_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12994_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500007057.mm.1 NM_007786.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007057.mm.1 NM_007786.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007057.mm.1 TC0500000802.mm.1 MTA TC
          Mouse Expression Set 430 1419735_PM_at - -
          Mouse Expression Set 430 1451851_PM_a_at - -
          Mouse Expression Set 430 1451851_a_at - -
          Mouse Expression Set 430 1419735_at - -
          Murine Genome U74v2 Set 99065_PM_at - -
          Murine Genome U74v2 Set 99065_at - -
          Mouse Genome 430A 2.0 Array 1419735_PM_at - -
          Mouse Genome 430A 2.0 Array 1451851_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1451851_a_at - -
          Mouse Genome 430A 2.0 Array 1419735_at - -
          Affymetrix HT Mouse Genome 430A Array 1419735_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419735_at - -
          Affymetrix HT Mouse Genome 430A Array 1451851_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451851_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 779084 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102060520 - -
          Illumina mouse-6 v1.0 expression beadchip 2340332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621622 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621622 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227118 - -
          mouse-6 v1.1 (Illumina) scl012994.5_66-S - -
          Illumina Sentrix mouse-6.v1 bead array 102060520 - -
          Illumina Sentrix mouse-6.v1 bead array 2340332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl012994.5_66-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102060520 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2854811 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621622 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227118 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2854811 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline